Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
SSMEM1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SSMEM1 - serine rich single-pass membrane protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145268.3 | 45 | Intron | NP_660311.1 | |||
XM_005250141.3 | 45 | Intron | XP_005250198.1 | |||
XM_011515794.2 | 45 | Intron | XP_011514096.1 | |||
XM_011515795.2 | 45 | UTR 5 | XP_011514097.1 |
TMEM209 - transmembrane protein 209 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301163.1 | 45 | UTR 5 | NP_001288092.1 | |||
NM_032842.3 | 45 | UTR 5 | NP_116231.2 | |||
XM_005250655.1 | 45 | Intron | XP_005250712.1 | |||
XM_011516646.2 | 45 | Intron | XP_011514948.1 |