Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300561 MIM: 300565 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLITRK2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SLITRK2 - SLIT and NTRK like family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144003.2 | 1186 | Intron | NP_001137475.1 | |||
NM_001144004.2 | 1186 | Intron | NP_001137476.1 | |||
NM_001144005.2 | 1186 | Intron | NP_001137477.1 | |||
NM_001144006.2 | 1186 | Intron | NP_001137478.1 | |||
NM_001144008.2 | 1186 | Intron | NP_001137480.1 | |||
NM_001144009.2 | 1186 | Intron | NP_001137481.1 | |||
NM_001144010.2 | 1186 | Intron | NP_001137482.1 | |||
NM_032539.4 | 1186 | Intron | NP_115928.1 | |||
XM_005262342.3 | 1186 | UTR 3 | XP_005262399.1 | |||
XM_005262343.3 | 1186 | UTR 3 | XP_005262400.1 | |||
XM_005262344.3 | 1186 | UTR 3 | XP_005262401.1 | |||
XM_005262345.3 | 1186 | UTR 3 | XP_005262402.1 |
TMEM257 - transmembrane protein 257 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004709.2 | 1186 | UTR 3 | NP_004700.1 |