Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 120105 MIM: 609427 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLPS PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CLPS - colipase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252597.1 | 79 | UTR 5 | NP_001239526.1 | |||
NM_001252598.1 | 79 | Missense Mutation | CCG,CTG | P,L 8 | NP_001239527.1 | |
NM_001832.3 | 79 | Missense Mutation | CCG,CTG | P,L 8 | NP_001823.1 |
CLPSL1 - colipase like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LHFPL5 - lipoma HMGIC fusion partner-like 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |