Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGAAGGTCACTAGGTGAAGAACCT[C/T]CATGAGGGAATGGTACTCCAGCCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604197 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MTHFS PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MTHFS - 5,10-methenyltetrahydrofolate synthetase (5-formyltetrahydrofolate cyclo-ligase) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199758.1 | Intron | NP_001186687.1 | ||||
NM_006441.3 | Intron | NP_006432.1 |
ST20-MTHFS - ST20-MTHFS readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199760.1 | Intron | NP_001186689.1 |
Set Membership: |
HapMap |