Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611627 MIM: 606969 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM57A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FAM57A - family with sequence similarity 57 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318006.1 | Intron | NP_001304935.1 | ||||
NM_001318007.1 | Intron | NP_001304936.1 | ||||
NM_001318008.1 | Intron | NP_001304937.1 | ||||
NM_024792.2 | Intron | NP_079068.1 |
GEMIN4 - gem nuclear organelle associated protein 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |