Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611592 MIM: 613311 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FARS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FARS2 - phenylalanyl-tRNA synthetase 2, mitochondrial | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318872.1 | 612 | Intron | NP_001305801.1 | |||
NM_006567.4 | 612 | UTR 5 | NP_006558.1 | |||
XM_005248812.3 | 612 | UTR 5 | XP_005248869.1 | |||
XM_006714966.2 | 612 | UTR 5 | XP_006715029.1 | |||
XM_011514247.2 | 612 | UTR 5 | XP_011512549.1 | |||
XM_011514248.2 | 612 | UTR 5 | XP_011512550.1 | |||
XM_011514249.1 | 612 | UTR 5 | XP_011512551.1 | |||
XM_011514251.2 | 612 | UTR 5 | XP_011512553.1 | |||
XM_017010186.1 | 612 | UTR 5 | XP_016865675.1 | |||
XM_017010187.1 | 612 | UTR 5 | XP_016865676.1 |
LYRM4 - LYR motif containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164840.2 | 612 | Intron | NP_001158312.1 | |||
NM_001164841.2 | 612 | Intron | NP_001158313.1 | |||
NM_001318782.1 | 612 | Intron | NP_001305711.1 | |||
NM_001318783.1 | 612 | Intron | NP_001305712.1 | |||
NM_020408.5 | 612 | Intron | NP_065141.3 | |||
XM_017011083.1 | 612 | Intron | XP_016866572.1 | |||
XM_017011084.1 | 612 | Intron | XP_016866573.1 |
Set Membership: |
HapMap |