Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602422 MIM: 605303 MIM: 615975 | ||||||||||||||||||||
Literature Links: |
SLBP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLBP - stem-loop binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001306074.1 | 2614 | Intron | NP_001293003.1 | |||
NM_001306075.1 | 2614 | Intron | NP_001293004.1 | |||
NM_006527.3 | 2614 | Intron | NP_006518.1 |
TACC3 - transforming acidic coiled-coil containing protein 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM129 - transmembrane protein 129 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127266.1 | 2614 | UTR 3 | NP_001120738.1 | |||
NM_138385.3 | 2614 | UTR 3 | NP_612394.1 | |||
XM_005248038.4 | 2614 | UTR 3 | XP_005248095.1 | |||
XM_005248039.4 | 2614 | UTR 3 | XP_005248096.1 | |||
XM_011513594.2 | 2614 | UTR 3 | XP_011511896.1 | |||
XM_017008834.1 | 2614 | UTR 3 | XP_016864323.1 |