Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCAACAGCAGCAGAGAGCTCGAAT[A/G]GCACCCTCTACACCACCACAGCTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
FRMPD3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FRMPD3 - FERM and PDZ domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032428.1 | Intron | NP_115804.1 | ||||
XM_006724709.2 | Intron | XP_006724772.1 | ||||
XM_011531056.2 | Intron | XP_011529358.1 | ||||
XM_011531057.2 | Intron | XP_011529359.1 | ||||
XM_011531058.2 | Intron | XP_011529360.1 | ||||
XM_017029901.1 | Intron | XP_016885390.1 | ||||
XM_017029902.1 | Intron | XP_016885391.1 |
FRMPD3-AS1 - FRMPD3 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |