Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176261 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCNE1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
KCNE1 - potassium voltage-gated channel subfamily E regulatory subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000219.5 | Intron | NP_000210.2 | ||||
NM_001127668.3 | Intron | NP_001121140.1 | ||||
NM_001127669.3 | Intron | NP_001121141.1 | ||||
NM_001127670.3 | Intron | NP_001121142.1 | ||||
NM_001270402.2 | Intron | NP_001257331.1 | ||||
NM_001270403.2 | Intron | NP_001257332.1 | ||||
NM_001270404.2 | Intron | NP_001257333.1 | ||||
NM_001270405.2 | Intron | NP_001257334.1 | ||||
XM_017028342.1 | Intron | XP_016883831.1 |
Set Membership: |
HapMap |