Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
42 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 146910 MIM: 147070 MIM: 147010 MIM: 147170 MIM: 147020 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IGH PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IGH - immunoglobulin heavy locus | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
IGHD - immunoglobulin heavy constant delta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
IGHM - immunoglobulin heavy constant mu | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR4507 - microRNA 4507 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR4537 - microRNA 4537 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR4538 - microRNA 4538 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR4539 - microRNA 4539 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |