Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 176741 | ||||||||||||||||||||
Literature Links: |
MKI67 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MKI67 - marker of proliferation Ki-67 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145966.1 | 7833 | Missense Mutation | ACA,AGA | T,R 2886 | NP_001139438.1 | |
NM_002417.4 | 7833 | Missense Mutation | ACA,AGA | T,R 3246 | NP_002408.3 | |
XM_006717864.3 | 7833 | Missense Mutation | ACA,AGA | T,R 2472 | XP_006717927.2 | |
XM_011539818.2 | 7833 | Missense Mutation | ACA,AGA | T,R 2902 | XP_011538120.1 |