Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 137150 MIM: 615261 | ||||||||||||||||||||
Literature Links: |
ABAT PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ABAT - 4-aminobutyrate aminotransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000663.4 | Intron | NP_000654.2 | ||||
NM_001127448.1 | Intron | NP_001120920.1 | ||||
NM_020686.5 | Intron | NP_065737.2 | ||||
XM_011522400.2 | Intron | XP_011520702.1 | ||||
XM_011522401.2 | Intron | XP_011520703.1 |
METTL22 - methyltransferase like 22 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |