Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608349 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HSH2D PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
HSH2D - hematopoietic SH2 domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291274.1 | Intron | NP_001278203.1 | ||||
NM_032855.3 | Intron | NP_116244.1 | ||||
XM_011528386.2 | Intron | XP_011526688.1 | ||||
XM_011528387.2 | Intron | XP_011526689.1 | ||||
XM_011528388.2 | Intron | XP_011526690.1 | ||||
XM_011528389.2 | Intron | XP_011526691.1 | ||||
XM_011528390.1 | Intron | XP_011526692.1 | ||||
XM_017027402.1 | Intron | XP_016882891.1 | ||||
XM_017027403.1 | Intron | XP_016882892.1 | ||||
XM_017027404.1 | Intron | XP_016882893.1 |
Set Membership: |
HapMap |