Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCCGAGCACACGGCTGAGCCCGGT[A/T]GGCAAAGGAAGGAAGGTCCAGGTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 138300 MIM: 602155 | ||||||||||||||||||||
Literature Links: |
GSR PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GSR - glutathione-disulfide reductase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC105379356 - uncharacterized LOC105379356 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_017014155.1 | 1109 | Intron | XP_016869644.1 | |||
XM_017014156.1 | 1109 | UTR 3 | XP_016869645.1 |
UBXN8 - UBX domain protein 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282189.1 | 1109 | Intron | NP_001269118.1 | |||
NM_001282199.1 | 1109 | Intron | NP_001269128.1 | |||
NM_005671.3 | 1109 | Intron | NP_005662.1 | |||
XM_011544655.2 | 1109 | Intron | XP_011542957.1 | |||
XM_017013862.1 | 1109 | Intron | XP_016869351.1 |