Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602879 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EPB41L1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
EPB41L1 - erythrocyte membrane protein band 4.1 like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258329.1 | Intron | NP_001245258.1 | ||||
NM_001258330.1 | Intron | NP_001245259.1 | ||||
NM_001258331.1 | Intron | NP_001245260.1 | ||||
NM_012156.2 | Intron | NP_036288.2 | ||||
NM_177996.2 | Intron | NP_818932.1 | ||||
XM_011528667.1 | Intron | XP_011526969.1 | ||||
XM_011528671.1 | Intron | XP_011526973.1 | ||||
XM_011528673.1 | Intron | XP_011526975.1 | ||||
XM_011528677.1 | Intron | XP_011526979.1 | ||||
XM_011528681.1 | Intron | XP_011526983.1 | ||||
XM_011528682.1 | Intron | XP_011526984.1 | ||||
XM_011528683.1 | Intron | XP_011526985.1 | ||||
XM_011528685.1 | Intron | XP_011526987.1 | ||||
XM_017027711.1 | Intron | XP_016883200.1 | ||||
XM_017027712.1 | Intron | XP_016883201.1 | ||||
XM_017027713.1 | Intron | XP_016883202.1 | ||||
XM_017027714.1 | Intron | XP_016883203.1 | ||||
XM_017027715.1 | Intron | XP_016883204.1 | ||||
XM_017027716.1 | Intron | XP_016883205.1 | ||||
XM_017027717.1 | Intron | XP_016883206.1 | ||||
XM_017027718.1 | Intron | XP_016883207.1 | ||||
XM_017027719.1 | Intron | XP_016883208.1 | ||||
XM_017027720.1 | Intron | XP_016883209.1 |