Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605654 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBXL4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FBXL4 - F-box and leucine rich repeat protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278716.1 | 6250 | Intron | NP_001265645.1 | |||
NM_012160.4 | 6250 | Intron | NP_036292.2 | |||
XM_005266930.2 | 6250 | UTR 3 | XP_005266987.1 | |||
XM_011535748.2 | 6250 | Intron | XP_011534050.1 | |||
XM_017010726.1 | 6250 | Intron | XP_016866215.1 | |||
XM_017010727.1 | 6250 | UTR 3 | XP_016866216.1 | |||
XM_017010728.1 | 6250 | UTR 3 | XP_016866217.1 |