Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTATGTTTTAAATATACTTTAAGGG[G/T]AAAAGTTGACTACCTCTCGATATTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 609287 | ||||||||||||||||||||
Literature Links: |
LOC105378828 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC105378828 - uncharacterized LOC105378828 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SH3GLB1 - SH3 domain containing GRB2 like endophilin B1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206651.1 | Intron | NP_001193580.1 | ||||
NM_001206652.1 | Intron | NP_001193581.1 | ||||
NM_001206653.1 | Intron | NP_001193582.1 | ||||
NM_016009.4 | Intron | NP_057093.1 | ||||
XM_006710672.2 | Intron | XP_006710735.1 | ||||
XM_006710673.2 | Intron | XP_006710736.1 | ||||
XM_011541538.1 | Intron | XP_011539840.1 | ||||
XM_017001408.1 | Intron | XP_016856897.1 | ||||
XM_017001409.1 | Intron | XP_016856898.1 | ||||
XM_017001410.1 | Intron | XP_016856899.1 |