Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603151 | ||||||||||||||||||||
Literature Links: |
SEPT7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SEPT7 - septin 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001011553.3 | Intron | NP_001011553.2 | ||||
NM_001242956.1 | Intron | NP_001229885.1 | ||||
NM_001788.5 | Intron | NP_001779.3 | ||||
XM_006715806.3 | Intron | XP_006715869.1 | ||||
XM_011515656.2 | Intron | XP_011513958.1 | ||||
XM_011515661.2 | Intron | XP_011513963.1 | ||||
XM_017012855.1 | Intron | XP_016868344.1 | ||||
XM_017012856.1 | Intron | XP_016868345.1 | ||||
XM_017012857.1 | Intron | XP_016868346.1 | ||||
XM_017012858.1 | Intron | XP_016868347.1 | ||||
XM_017012859.1 | Intron | XP_016868348.1 | ||||
XM_017012860.1 | Intron | XP_016868349.1 | ||||
XM_017012861.1 | Intron | XP_016868350.1 | ||||
XM_017012862.1 | Intron | XP_016868351.1 | ||||
XM_017012863.1 | Intron | XP_016868352.1 |
SEPT7-AS1 - SEPT7 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |