Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614242 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC16A9 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
SLC16A9 - solute carrier family 16 member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001323977.1 | 2770 | UTR 3 | NP_001310906.1 | |||
NM_001323978.1 | 2770 | UTR 3 | NP_001310907.1 | |||
NM_001323979.1 | 2770 | UTR 3 | NP_001310908.1 | |||
NM_001323980.1 | 2770 | UTR 3 | NP_001310909.1 | |||
NM_001323981.1 | 2770 | UTR 3 | NP_001310910.1 | |||
NM_194298.2 | 2770 | UTR 3 | NP_919274.1 | |||
XM_017015883.1 | 2770 | UTR 3 | XP_016871372.1 | |||
XM_017015884.1 | 2770 | UTR 3 | XP_016871373.1 |
Set Membership: |
HapMap |