Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
38 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608449 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTBP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PTBP2 - polypyrimidine tract binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300985.1 | Intron | NP_001287914.1 | ||||
NM_001300986.1 | Intron | NP_001287915.1 | ||||
NM_001300987.1 | Intron | NP_001287916.1 | ||||
NM_001300988.1 | Intron | NP_001287917.1 | ||||
NM_001300989.1 | Intron | NP_001287918.1 | ||||
NM_001300990.1 | Intron | NP_001287919.1 | ||||
NM_021190.3 | Intron | NP_067013.1 | ||||
XM_011541874.2 | Intron | XP_011540176.1 | ||||
XM_011541875.2 | Intron | XP_011540177.1 | ||||
XM_011541876.2 | Intron | XP_011540178.1 | ||||
XM_017001951.1 | Intron | XP_016857440.1 |