Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615690 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AMIGO2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AMIGO2 - adhesion molecule with Ig like domain 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143668.1 | Intron | NP_001137140.1 | ||||
NM_181847.4 | Intron | NP_862830.1 | ||||
XM_005268837.3 | Intron | XP_005268894.1 |
PCED1B - PC-esterase domain containing 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281429.1 | Intron | NP_001268358.1 | ||||
NM_138371.2 | Intron | NP_612380.1 | ||||
XM_005269224.4 | Intron | XP_005269281.1 | ||||
XM_011538978.2 | Intron | XP_011537280.1 | ||||
XM_017020207.1 | Intron | XP_016875696.1 | ||||
XM_017020208.1 | Intron | XP_016875697.1 | ||||
XM_017020209.1 | Intron | XP_016875698.1 | ||||
XM_017020210.1 | Intron | XP_016875699.1 | ||||
XM_017020211.1 | Intron | XP_016875700.1 | ||||
XM_017020212.1 | Intron | XP_016875701.1 | ||||
XM_017020213.1 | Intron | XP_016875702.1 | ||||
XM_017020214.1 | Intron | XP_016875703.1 | ||||
XM_017020215.1 | Intron | XP_016875704.1 | ||||
XM_017020216.1 | Intron | XP_016875705.1 |