Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
55 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610171 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CALML6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CALML6 - calmodulin like 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CFAP74 - cilia and flagella associated protein 74 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304360.1 | 4322 | Silent Mutation | TCC,TCT | S,S 1579 | NP_001291289.1 | |
XM_011542333.2 | 4322 | Silent Mutation | TCC,TCT | S,S 1602 | XP_011540635.2 | |
XM_011542335.1 | 4322 | Silent Mutation | TCC,TCT | S,S 1410 | XP_011540637.1 | |
XM_011542336.2 | 4322 | Silent Mutation | TCC,TCT | S,S 1410 | XP_011540638.1 | |
XM_017002641.1 | 4322 | Silent Mutation | TCC,TCT | S,S 1621 | XP_016858130.1 | |
XM_017002642.1 | 4322 | Silent Mutation | TCC,TCT | S,S 1611 | XP_016858131.1 | |
XM_017002643.1 | 4322 | Silent Mutation | TCC,TCT | S,S 1621 | XP_016858132.1 | |
XM_017002644.1 | 4322 | Silent Mutation | TCC,TCT | S,S 765 | XP_016858133.1 |
TMEM52 - transmembrane protein 52 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |