Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609844 MIM: 159350 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DCP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DCP2 - decapping mRNA 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242377.1 | 7626 | Intron | NP_001229306.1 | |||
NM_152624.5 | 7626 | Intron | NP_689837.2 | |||
XM_017009163.1 | 7626 | Intron | XP_016864652.1 |
MCC - mutated in colorectal cancers | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001085377.1 | 7626 | UTR 3 | NP_001078846.1 | |||
NM_002387.2 | 7626 | UTR 3 | NP_002378.1 | |||
XM_005271991.2 | 7626 | UTR 3 | XP_005272048.1 | |||
XM_017009473.1 | 7626 | UTR 3 | XP_016864962.1 | |||
XM_017009474.1 | 7626 | UTR 3 | XP_016864963.1 |