Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607394 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC649133 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC649133 - uncharacterized LOC649133 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
POU2F3 - POU class 2 homeobox 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001244682.1 | Intron | NP_001231611.1 | ||||
NM_014352.3 | Intron | NP_055167.2 | ||||
XM_011542739.2 | Intron | XP_011541041.1 | ||||
XM_011542740.2 | Intron | XP_011541042.1 | ||||
XM_011542741.2 | Intron | XP_011541043.1 | ||||
XM_011542742.2 | Intron | XP_011541044.1 | ||||
XM_011542743.2 | Intron | XP_011541045.1 | ||||
XM_017017487.1 | Intron | XP_016872976.1 |