Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607394 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC649133 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC649133 - uncharacterized LOC649133 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
POU2F3 - POU class 2 homeobox 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001244682.1 | 1296 | Intron | NP_001231611.1 | |||
NM_014352.3 | 1296 | Intron | NP_055167.2 | |||
XM_011542739.2 | 1296 | Intron | XP_011541041.1 | |||
XM_011542740.2 | 1296 | Intron | XP_011541042.1 | |||
XM_011542741.2 | 1296 | Intron | XP_011541043.1 | |||
XM_011542742.2 | 1296 | Intron | XP_011541044.1 | |||
XM_011542743.2 | 1296 | UTR 5 | XP_011541045.1 | |||
XM_017017487.1 | 1296 | Intron | XP_016872976.1 |