Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608080 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MRO PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MRO - maestro | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127174.1 | 2699 | UTR 3 | NP_001120646.1 | |||
NM_001127175.1 | 2699 | UTR 3 | NP_001120647.1 | |||
NM_001127176.1 | 2699 | UTR 3 | NP_001120648.1 | |||
NM_031939.3 | 2699 | UTR 3 | NP_114145.2 | |||
XM_005258365.3 | 2699 | UTR 3 | XP_005258422.1 | |||
XM_011526226.2 | 2699 | UTR 3 | XP_011524528.1 | |||
XM_017026035.1 | 2699 | UTR 3 | XP_016881524.1 | |||
XM_017026036.1 | 2699 | UTR 3 | XP_016881525.1 | |||
XM_017026037.1 | 2699 | UTR 3 | XP_016881526.1 |