Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612134 | ||||||||||||||||||||
Literature Links: |
GLCE PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GLCE - glucuronic acid epimerase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324091.1 | Intron | NP_001311020.1 | ||||
NM_001324092.1 | Intron | NP_001311021.1 | ||||
NM_001324093.1 | Intron | NP_001311022.1 | ||||
NM_001324094.1 | Intron | NP_001311023.1 | ||||
NM_015554.2 | Intron | NP_056369.1 | ||||
XM_005254298.3 | Intron | XP_005254355.1 | ||||
XM_017022073.1 | Intron | XP_016877562.1 | ||||
XM_017022074.1 | Intron | XP_016877563.1 |