Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGCCTTACAGCTGCCCAGCTATT[G/T]CCCTAATCTCAAGCCCCTTTACTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
23 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 179520 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC107985184 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC107985184 - protein enabled homolog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RAP1A - RAP1A, member of RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010935.2 | Intron | NP_001010935.1 | ||||
NM_001291896.1 | Intron | NP_001278825.1 | ||||
NM_002884.3 | Intron | NP_002875.1 | ||||
XM_017001960.1 | Intron | XP_016857449.1 | ||||
XM_017001961.1 | Intron | XP_016857450.1 | ||||
XM_017001962.1 | Intron | XP_016857451.1 | ||||
XM_017001963.1 | Intron | XP_016857452.1 | ||||
XM_017001964.1 | Intron | XP_016857453.1 |
Set Membership: |
HapMap |