Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
49 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607020 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RASSF5 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
RASSF5 - Ras association domain family member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182663.3 | 121 | Silent Mutation | CTA,CTC | L,L 16 | NP_872604.1 | |
NM_182664.3 | 121 | Silent Mutation | CTA,CTC | L,L 16 | NP_872605.1 | |
NM_182665.3 | 121 | Intron | NP_872606.1 |
Set Membership: |
HapMap |