Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTCTTAGATATGACATCAAAAGC[A/T]CAGTCAATTAAAAAAATGAACTTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602240 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZKSCAN8 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZKSCAN8 - zinc finger with KRAB and SCAN domains 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278119.1 | Intron | NP_001265048.1 | ||||
NM_001278121.1 | Intron | NP_001265050.1 | ||||
NM_001278122.1 | Intron | NP_001265051.1 | ||||
NM_006298.3 | Intron | NP_006289.2 | ||||
XM_011514870.2 | Intron | XP_011513172.1 | ||||
XM_011514873.2 | Intron | XP_011513175.1 | ||||
XM_011514874.2 | Intron | XP_011513176.1 | ||||
XM_017011266.1 | Intron | XP_016866755.1 |
ZSCAN16-AS1 - ZSCAN16 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |