Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611343 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IDNK PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IDNK - IDNK, gluconokinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001551.3 | Intron | NP_001001551.2 | ||||
NM_001256915.1 | Intron | NP_001243844.1 | ||||
XM_006717110.3 | Intron | XP_006717173.1 | ||||
XM_006717111.3 | Intron | XP_006717174.1 | ||||
XM_017014719.1 | Intron | XP_016870208.1 | ||||
XM_017014720.1 | Intron | XP_016870209.1 | ||||
XM_017014721.1 | Intron | XP_016870210.1 | ||||
XM_017014722.1 | Intron | XP_016870211.1 | ||||
XM_017014723.1 | Intron | XP_016870212.1 | ||||
XM_017014724.1 | Intron | XP_016870213.1 | ||||
XM_017014725.1 | Intron | XP_016870214.1 |