Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604090 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DLG5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
DLG5 - discs large MAGUK scaffold protein 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004747.3 | 5331 | UTR 3 | NP_004738.3 | |||
XM_005270276.4 | 5331 | UTR 3 | XP_005270333.1 | |||
XM_006718056.3 | 5331 | UTR 3 | XP_006718119.1 | |||
XM_006718057.3 | 5331 | Intron | XP_006718120.1 | |||
XM_011540341.2 | 5331 | UTR 3 | XP_011538643.1 | |||
XM_011540342.1 | 5331 | UTR 3 | XP_011538644.1 | |||
XM_011540344.2 | 5331 | UTR 3 | XP_011538646.1 | |||
XM_011540345.1 | 5331 | UTR 3 | XP_011538647.1 | |||
XM_011540346.2 | 5331 | Intron | XP_011538648.1 | |||
XM_011540347.2 | 5331 | UTR 3 | XP_011538649.1 | |||
XM_017016913.1 | 5331 | UTR 3 | XP_016872402.1 | |||
XM_017016914.1 | 5331 | UTR 3 | XP_016872403.1 | |||
XM_017016915.1 | 5331 | UTR 3 | XP_016872404.1 |
Set Membership: |
HapMap Validated |