Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACCCTCCTCTTAGAGGGGCGAGTG[A/G]CCTTTATGCCAGGTTCTGCTAAAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
24 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613514 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100130331 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC100130331 - POTE ankyrin domain family, member F pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZP4 - zona pellucida glycoprotein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021186.4 | Intron | NP_067009.1 |