Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604483 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SIRT5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SIRT5 - sirtuin 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193267.2 | 86 | Intron | NP_001180196.1 | |||
NM_001242827.1 | 86 | Intron | NP_001229756.1 | |||
NM_012241.4 | 86 | UTR 5 | NP_036373.1 | |||
NM_031244.3 | 86 | Intron | NP_112534.1 | |||
XM_005248968.4 | 86 | Intron | XP_005249025.1 | |||
XM_011514444.2 | 86 | Intron | XP_011512746.1 | |||
XM_017010620.1 | 86 | Intron | XP_016866109.1 | |||
XM_017010621.1 | 86 | Intron | XP_016866110.1 | |||
XM_017010622.1 | 86 | Intron | XP_016866111.1 | |||
XM_017010623.1 | 86 | UTR 5 | XP_016866112.1 | |||
XM_017010624.1 | 86 | Intron | XP_016866113.1 | |||
XM_017010625.1 | 86 | Intron | XP_016866114.1 |