Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603889 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SIRPB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SIRPB1 - signal regulatory protein beta 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005260641.3 | Intron | XP_005260698.1 | ||||
XM_005260643.3 | Intron | XP_005260700.1 | ||||
XM_005260644.3 | Intron | XP_005260701.1 | ||||
XM_011529130.2 | Intron | XP_011527432.1 | ||||
XM_011529134.2 | Intron | XP_011527436.1 | ||||
XM_017027577.1 | Intron | XP_016883066.1 | ||||
XM_017027578.1 | Intron | XP_016883067.1 | ||||
XM_017027579.1 | Intron | XP_016883068.1 | ||||
XM_017027580.1 | Intron | XP_016883069.1 | ||||
XM_017027581.1 | Intron | XP_016883070.1 | ||||
XM_017027582.1 | Intron | XP_016883071.1 | ||||
XM_017027583.1 | Intron | XP_016883072.1 | ||||
XM_017027584.1 | Intron | XP_016883073.1 | ||||
XM_017027585.1 | Intron | XP_016883074.1 |
Set Membership: |
HapMap |