Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 114206 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CACNA1D PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CACNA1D - calcium voltage-gated channel subunit alpha1 D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000720.3 | Intron | NP_000711.1 | ||||
NM_001128839.2 | Intron | NP_001122311.1 | ||||
NM_001128840.2 | Intron | NP_001122312.1 | ||||
XM_005265448.3 | Intron | XP_005265505.1 | ||||
XM_011534094.2 | Intron | XP_011532396.1 | ||||
XM_011534096.2 | Intron | XP_011532398.1 | ||||
XM_011534097.2 | Intron | XP_011532399.1 | ||||
XM_011534099.2 | Intron | XP_011532401.1 | ||||
XM_011534100.2 | Intron | XP_011532402.1 | ||||
XM_017007137.1 | Intron | XP_016862626.1 | ||||
XM_017007138.1 | Intron | XP_016862627.1 | ||||
XM_017007139.1 | Intron | XP_016862628.1 | ||||
XM_017007140.1 | Intron | XP_016862629.1 | ||||
XM_017007141.1 | Intron | XP_016862630.1 | ||||
XM_017007142.1 | Intron | XP_016862631.1 | ||||
XM_017007143.1 | Intron | XP_016862632.1 | ||||
XM_017007144.1 | Intron | XP_016862633.1 | ||||
XM_017007145.1 | Intron | XP_016862634.1 |