Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607978 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SAMSN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SAMSN1 - SAM domain, SH3 domain and nuclear localization signals 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256370.1 | Intron | NP_001243299.1 | ||||
NM_001286523.1 | Intron | NP_001273452.1 | ||||
NM_022136.4 | Intron | NP_071419.3 | ||||
XM_011529684.2 | Intron | XP_011527986.1 | ||||
XM_011529685.1 | Intron | XP_011527987.1 | ||||
XM_011529686.1 | Intron | XP_011527988.1 | ||||
XM_017028427.1 | Intron | XP_016883916.1 |