Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611163 | ||||||||||||||||||||
Literature Links: |
TOX2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TOX2 - TOX high mobility group box family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098796.1 | Intron | NP_001092266.1 | ||||
NM_001098797.1 | Intron | NP_001092267.1 | ||||
NM_001098798.1 | Intron | NP_001092268.1 | ||||
NM_032883.2 | Intron | NP_116272.1 | ||||
XM_006723884.1 | Intron | XP_006723947.1 | ||||
XM_017028109.1 | Intron | XP_016883598.1 |