Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607781 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PAQR5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PAQR5 - progestin and adipoQ receptor family member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001104554.1 | Intron | NP_001098024.1 | ||||
NM_017705.3 | Intron | NP_060175.3 | ||||
XM_005254495.4 | Intron | XP_005254552.1 | ||||
XM_011521720.2 | Intron | XP_011520022.1 | ||||
XM_011521722.2 | Intron | XP_011520024.1 | ||||
XM_011521723.2 | Intron | XP_011520025.1 | ||||
XM_011521725.2 | Intron | XP_011520027.1 | ||||
XM_011521727.2 | Intron | XP_011520029.1 | ||||
XM_017022360.1 | Intron | XP_016877849.1 | ||||
XM_017022361.1 | Intron | XP_016877850.1 | ||||
XM_017022362.1 | Intron | XP_016877851.1 | ||||
XM_017022363.1 | Intron | XP_016877852.1 | ||||
XM_017022364.1 | Intron | XP_016877853.1 | ||||
XM_017022365.1 | Intron | XP_016877854.1 | ||||
XM_017022366.1 | Intron | XP_016877855.1 | ||||
XM_017022367.1 | Intron | XP_016877856.1 | ||||
XM_017022368.1 | Intron | XP_016877857.1 |
Set Membership: |
HapMap |