Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606645 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARAP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ARAP2 - ArfGAP with RhoGAP domain, ankyrin repeat and PH domain 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015230.3 | Intron | NP_056045.2 | ||||
XM_017007698.1 | Intron | XP_016863187.1 | ||||
XM_017007699.1 | Intron | XP_016863188.1 | ||||
XM_017007700.1 | Intron | XP_016863189.1 | ||||
XM_017007701.1 | Intron | XP_016863190.1 | ||||
XM_017007702.1 | Intron | XP_016863191.1 |
Set Membership: |
HapMap |