Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613221 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MTCH2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MTCH2 - mitochondrial carrier 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317231.1 | Intron | NP_001304160.1 | ||||
NM_001317232.1 | Intron | NP_001304161.1 | ||||
NM_001317233.1 | Intron | NP_001304162.1 | ||||
NM_014342.3 | Intron | NP_055157.1 | ||||
XM_006718172.2 | Intron | XP_006718235.1 | ||||
XM_011519959.2 | Intron | XP_011518261.1 | ||||
XM_011519960.2 | Intron | XP_011518262.1 | ||||
XM_011519961.2 | Intron | XP_011518263.1 | ||||
XM_017017462.1 | Intron | XP_016872951.1 | ||||
XM_017017463.1 | Intron | XP_016872952.1 |
Set Membership: |
HapMap |