Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604066 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
B3GALT5 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
B3GALT5 - beta-1,3-galactosyltransferase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278650.1 | Intron | NP_001265579.1 | ||||
NM_006057.2 | Intron | NP_006048.1 | ||||
NM_033170.2 | Intron | NP_149360.1 | ||||
NM_033171.2 | Intron | NP_149361.1 | ||||
NM_033172.2 | Intron | NP_149362.2 | ||||
XM_017028227.1 | Intron | XP_016883716.1 | ||||
XM_017028228.1 | Intron | XP_016883717.1 | ||||
XM_017028229.1 | Intron | XP_016883718.1 | ||||
XM_017028230.1 | Intron | XP_016883719.1 | ||||
XM_017028231.1 | Intron | XP_016883720.1 | ||||
XM_017028232.1 | Intron | XP_016883721.1 | ||||
XM_017028233.1 | Intron | XP_016883722.1 | ||||
XM_017028234.1 | Intron | XP_016883723.1 | ||||
XM_017028235.1 | Intron | XP_016883724.1 | ||||
XM_017028236.1 | Intron | XP_016883725.1 | ||||
XM_017028237.1 | Intron | XP_016883726.1 | ||||
XM_017028238.1 | Intron | XP_016883727.1 | ||||
XM_017028239.1 | Intron | XP_016883728.1 | ||||
XM_017028240.1 | Intron | XP_016883729.1 | ||||
XM_017028241.1 | Intron | XP_016883730.1 |
Set Membership: |
HapMap |