Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604960 | ||||||||||||||||||||
Literature Links: |
PACSIN2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PACSIN2 - protein kinase C and casein kinase substrate in neurons 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184970.1 | Intron | NP_001171899.1 | ||||
NM_001184971.1 | Intron | NP_001171900.1 | ||||
NM_007229.3 | Intron | NP_009160.2 | ||||
XM_005261319.3 | Intron | XP_005261376.1 | ||||
XM_011529846.1 | Intron | XP_011528148.1 | ||||
XM_011529847.2 | Intron | XP_011528149.1 | ||||
XM_011529848.2 | Intron | XP_011528150.1 | ||||
XM_011529849.2 | Intron | XP_011528151.1 | ||||
XM_011529850.2 | Intron | XP_011528152.1 | ||||
XM_011529851.2 | Intron | XP_011528153.1 | ||||
XM_011529852.1 | Intron | XP_011528154.1 | ||||
XM_011529853.1 | Intron | XP_011528155.1 | ||||
XM_017028562.1 | Intron | XP_016884051.1 | ||||
XM_017028563.1 | Intron | XP_016884052.1 | ||||
XM_017028564.1 | Intron | XP_016884053.1 | ||||
XM_017028565.1 | Intron | XP_016884054.1 | ||||
XM_017028566.1 | Intron | XP_016884055.1 | ||||
XM_017028567.1 | Intron | XP_016884056.1 | ||||
XM_017028568.1 | Intron | XP_016884057.1 |