Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605333 | ||||||||||||||||||||
Literature Links: |
PDS5B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PDS5B - PDS5 cohesin associated factor B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015032.3 | Intron | NP_055847.1 | ||||
XM_005266298.4 | Intron | XP_005266355.1 | ||||
XM_011534999.2 | Intron | XP_011533301.1 | ||||
XM_011535000.2 | Intron | XP_011533302.1 | ||||
XM_011535002.2 | Intron | XP_011533304.1 | ||||
XM_017020448.1 | Intron | XP_016875937.1 | ||||
XM_017020449.1 | Intron | XP_016875938.1 | ||||
XM_017020450.1 | Intron | XP_016875939.1 | ||||
XM_017020451.1 | Intron | XP_016875940.1 | ||||
XM_017020452.1 | Intron | XP_016875941.1 | ||||
XM_017020453.1 | Intron | XP_016875942.1 |