Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGGCAGACCTTAGTTGGTATTCA[A/G]GTTTCTACTAATGTCTCCCCAGCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100133315 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
LOC100133315 - transient receptor potential cation channel, subfamily C, member 2-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNF121 - ring finger protein 121 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300926.1 | Intron | NP_001287855.1 | ||||
NM_018320.4 | Intron | NP_060790.2 | ||||
XM_006718629.1 | Intron | XP_006718692.1 | ||||
XM_006718630.1 | Intron | XP_006718693.1 | ||||
XM_006718632.1 | Intron | XP_006718695.1 | ||||
XM_011545150.1 | Intron | XP_011543452.1 |
Set Membership: |
HapMap Validated |