Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNOT6L PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CNOT6L - CCR4-NOT transcription complex subunit 6 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286790.1 | 6096 | UTR 3 | NP_001273719.1 | |||
NM_144571.2 | 6096 | UTR 3 | NP_653172.2 | |||
XM_011531806.2 | 6096 | UTR 3 | XP_011530108.1 | |||
XM_011531807.2 | 6096 | UTR 3 | XP_011530109.1 | |||
XM_011531808.2 | 6096 | UTR 3 | XP_011530110.1 | |||
XM_011531809.1 | 6096 | UTR 3 | XP_011530111.1 | |||
XM_011531810.1 | 6096 | UTR 3 | XP_011530112.1 | |||
XM_017007956.1 | 6096 | UTR 3 | XP_016863445.1 | |||
XM_017007957.1 | 6096 | UTR 3 | XP_016863446.1 |
Set Membership: |
HapMap |