Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604839 MIM: 612548 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FKBP6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FKBP6 - FK506 binding protein 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135211.2 | 510 | Missense Mutation | AAG,GAG | K,E 131 | NP_001128683.1 | |
NM_001281304.1 | 510 | Missense Mutation | AAG,GAG | K,E 106 | NP_001268233.1 | |
NM_003602.4 | 510 | Missense Mutation | AAG,GAG | K,E 136 | NP_003593.3 | |
XM_006716153.2 | 510 | Missense Mutation | AAG,GAG | K,E 131 | XP_006716216.1 | |
XM_017012741.1 | 510 | Missense Mutation | AAG,GAG | K,E 131 | XP_016868230.1 | |
XM_017012742.1 | 510 | Missense Mutation | AAG,GAG | K,E 131 | XP_016868231.1 |
TRIM50 - tripartite motif containing 50 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |