Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613570 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LDAH PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LDAH - lipid droplet associated hydrolase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282719.1 | 2551 | UTR 3 | NP_001269648.1 | |||
NM_001282720.1 | 2551 | UTR 3 | NP_001269649.1 | |||
NM_001282721.1 | 2551 | UTR 3 | NP_001269650.1 | |||
NM_001282722.1 | 2551 | UTR 3 | NP_001269651.1 | |||
NM_001282723.1 | 2551 | UTR 3 | NP_001269652.1 | |||
NM_001282724.1 | 2551 | UTR 3 | NP_001269653.1 | |||
NM_021925.3 | 2551 | UTR 3 | NP_068744.1 | |||
XM_011533014.2 | 2551 | UTR 3 | XP_011531316.1 |