Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610561 MIM: 608547 | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRSS53 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | |||||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
PRSS53 - protease, serine 53 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VKORC1 - vitamin K epoxide reductase complex subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001311311.1 | Intron | NP_001298240.1 | ||||
NM_024006.5 | Intron | NP_076869.1 | ||||
NM_206824.2 | Intron | NP_996560.1 |
Set Membership: |
HapMap DME Validated Inventoried |