Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 136510 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FPGS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FPGS - folylpolyglutamate synthase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001018078.2 | Intron | NP_001018088.1 | ||||
NM_001288803.1 | Intron | NP_001275732.1 | ||||
NM_004957.5 | Intron | NP_004948.4 | ||||
XM_005251864.3 | Intron | XP_005251921.1 | ||||
XM_011518439.2 | Intron | XP_011516741.1 | ||||
XM_017014565.1 | Intron | XP_016870054.1 | ||||
XM_017014566.1 | Intron | XP_016870055.1 |